Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01237
- Entrez Gene ID
-
101927289
- Gene Name
- long intergenic non-protein coding RNA 1237
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 241881363 - 242078722 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110220.1 |
|
1 - 2 |
122 |
98 |
GenBank mRNA |
BG772076.1 |
- |
1 - 2 |
127 |
98 |
DA781142.1 |
- |
1 - 2 |
119 |
98 |
DA937565.1 |
- |
1 - 2 |
118 |
98 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB124644.1 |
- |
1 - 2 |
118 |
|
Chr. 2 - 241881367 - 241966120 [+] on Build GRCh38 |
98 |
GenBankmRNA |
DA816995.1 |
- |
1 - 2 |
112 |
|
Chr. 2 - 241881373 - 241966308 [+] on Build GRCh38 |
98 |
GenBankmRNA |
DA813420.1 |
- |
1 - 2 |
118 |
|
Chr. 2 - 241881367 - 241966140 [+] on Build GRCh38 |
98 |
GenBankmRNA |
DB141815.1 |
- |
1 - 2 |
118 |
|
Chr. 2 - 241881367 - 241934127 [+] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
receptor transporter protein 5 (putative) |
RTP5 |
C2orf85, CXXC11, Z3CXXC5 |
-7.5kb
|
Sense |
Chr. - 241869729 - 241873817 on Build GRCh38 |
Downstream |
uncharacterized LOC285095 |
LOC285095 |
|
11.4kb
|
Anti-sense |
Chr. - 241893983 - 241902623 on Build GRCh38 |
View Full Product Details