Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP6V0E2
Entrez Gene ID
155066
Gene Name
ATPase H+ transporting V0 subunit e2
Gene Aliases
ATP6V0E2L, C7orf32
Chromosome Location
Chr.7: 149872968 - 149880713 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110612.1 4 - 5 1424 72
GenBank mRNA AK172725.1 - 4 - 5 405 72


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM818605.1 - 4 - 5 410 Chr. 7 - 149873982 - 149879512 [+] on Build GRCh38 72
GenBankmRNA BM818667.1 - 4 - 5 410 Chr. 7 - 149873982 - 149879498 [+] on Build GRCh38 72
GenBankmRNA DA731441.1 - 4 - 5 404 Chr. 7 - 149873987 - 149879736 [+] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ATP6V0E2 antisense RNA 1 ATP6V0E2-AS1 -5.3kb Anti-sense Chr. - 149867694 - 149873862 on Build GRCh38
Downstream ARP3 actin-related protein 3 homolog C ACTR3C ARP11 6.5kb Anti-sense Chr. - 149881359 - 150325523 on Build GRCh38

View Full Product Details