The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_110659.1 | 1 - 2 | 96 | 104 | |
GenBank mRNA | AW161363.1 | - | 1 - 2 | 96 | 104 |
DR033788.1 | - | 1 - 2 | 144 | 104 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | poly(A) binding protein cytoplasmic 5 | PABPC5 | PABP5 | Anti-sense | Chr. - 91434598 - 91438584 on Build GRCh38 | |
Downstream | protocadherin 11 X-linked | PCDH11X | PCDH-X, PCDH11, PCDHX, PPP1R119 | 344.3kb | Anti-sense | Chr. - 91779310 - 92623230 on Build GRCh38 |