Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101929099
- Entrez Gene ID
-
101929099
- Gene Name
- uncharacterized LOC101929099
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 117025482 - 117059490 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110786.1 |
|
2 - 3 |
288 |
97 |
GenBank mRNA |
AK124329.1 |
- |
2 - 3 |
288 |
97 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB245993.1 |
- |
2 - 3 |
288 |
|
Chr. 1 - 117026719 - 117059490 [-] on Build GRCh38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
CD101 molecule |
CD101 |
EWI-101, IGSF2, V7 |
|
Anti-sense |
Chr. - 116990423 - 117037481 on Build GRCh38 |
Downstream |
transcription termination factor 2 |
TTF2 |
HuF2, ZGRF6 |
10.0kb
|
Anti-sense |
Chr. - 117060288 - 117107453 on Build GRCh38 |
View Full Product Details