Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101927814
- Entrez Gene ID
-
101927814
- Gene Name
- uncharacterized LOC101927814
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 21589698 - 21599385 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110930.1 |
|
1 - 2 |
729 |
83 |
GenBank mRNA |
BC036249.1 |
- |
1 - 2 |
729 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 7 member 5 pseudogene 2 |
SLC7A5P2 |
IMAA, MMAA |
-77.2kb
|
Sense |
Chr. - 21517893 - 21520444 on Build GRCh38 |
Downstream |
immunoglobulin superfamily member 6 |
IGSF6 |
DORA |
41.9kb
|
Sense |
Chr. - 21641284 - 21652666 on Build GRCh38 |
View Full Product Details