The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_111981.1 | 6 - 7 | 662 | 73 | |
GenBank mRNA | BC043383.1 | - | 6 - 7 | 662 | 73 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | SID1 transmembrane family member 1 | SIDT1 | SID-1, SID1 | -16.3kb | Anti-sense | Chr. - 113531774 - 113629579 on Build GRCh38 |
Downstream | N(alpha)-acetyltransferase 50, NatE catalytic subunit | NAA50 | MAK3, NAT13, NAT13P, NAT5, NAT5P, SAN | 52.1kb | Sense | Chr. - 113716460 - 113746299 on Build GRCh38 |