Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NMU
- Entrez Gene ID
-
10874
- Gene Name
- neuromedin U
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 55595229 - 55636698 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120489.1 |
|
5 - 6 |
355 |
77 |
GenBank mRNA |
AJ510134.1 |
- |
4 - 5 |
195 |
77 |
CN343322.1 |
- |
5 - 6 |
355 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
phosducin like 2 |
PDCL2 |
GCPHLP |
-13.0kb
|
Sense |
Chr. - 55556523 - 55592275 on Build GRCh38 |
Downstream |
exocyst complex component 1 pseudogene |
LOC644145 |
|
212.6kb
|
Anti-sense |
Chr. - 55820071 - 55837264 on Build GRCh38 |
View Full Product Details