Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC102724933
- Entrez Gene ID
-
102724933
- Gene Name
- uncharacterized LOC102724933
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 93174366 - 93181832 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120492.1 |
|
2 - 3 |
742 |
91 |
GenBank mRNA |
CB049231.1 |
- |
1 - 2 |
|
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC643339 |
LOC643339 |
|
|
Anti-sense |
Chr. - 93003758 - 93377736 on Build GRCh38 |
Downstream |
nudix hydrolase 4 |
NUDT4 |
DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P |
196.1kb
|
Sense |
Chr. - 93377923 - 93403248 on Build GRCh38 |
View Full Product Details