The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_120589.1 | 1 - 2 | 71 | 72 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | neuronal PAS domain protein 4 | NPAS4 | Le-PAS, NXF, PASD10, bHLHe79 | -11.5kb | Anti-sense | Chr. - 66409158 - 66426707 on Build GRCh38 |
Downstream | pellino E3 ubiquitin protein ligase family member 3 | PELI3 | 27.5kb | Anti-sense | Chr. - 66466327 - 66477337 on Build GRCh38 |