Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01315
- Entrez Gene ID
-
102723775
- Gene Name
- long intergenic non-protein coding RNA 1315
- Gene Aliases
-
lnc-C22orf32-1
- Chromosome Location
-
Chr.22: 42364400 - 42369208 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120595.1 |
|
1 - 2 |
496 |
69 |
GenBank mRNA |
BC038245.1 |
- |
1 - 2 |
462 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI915409.1 |
- |
1 - 2 |
462 |
|
Chr. 22 - 42364694 - 42369174 [-] on Build GRCh38 |
69 |
GenBankmRNA |
BF530005.1 |
- |
1 - 2 |
289 |
|
Chr. 22 - 42364640 - 42369000 [-] on Build GRCh38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
transcription factor 20 |
TCF20 |
AR1, SPBP, TCF-20 |
-80.5kb
|
Sense |
Chr. - 42160013 - 42283927 on Build GRCh38 |
Downstream |
NFAT activating protein with ITAM motif 1 |
NFAM1 |
CNAIP |
11.2kb
|
Sense |
Chr. - 42380407 - 42438155 on Build GRCh38 |
View Full Product Details