Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100996583
Entrez Gene ID
100996583
Gene Name
uncharacterized LOC100996583
Gene Aliases
-
Chromosome Location
Chr.1: 2566535 - 2584534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_121638.1 2 - 3 163 87
GenBank mRNA CB852498.1 - 2 - 3 87


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream TNF receptor superfamily member 14 TNFRSF14 ATAR, CD270, HVEA, HVEM, LIGHTR, TR2 -15.9kb Sense Chr. - 2555767 - 2565622 on Build GRCh38
Downstream family with sequence similarity 213 member B FAM213B C1orf93 3.0kb Sense Chr. - 2586460 - 2591469 on Build GRCh38

View Full Product Details