The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_125441.1 | 3 - 4 | 402 | 104 | |
GenBank mRNA | BC013846.1 | - | 3 - 4 | 322 | 104 |
DA433004.1 | - | 3 - 4 | 321 | 104 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | prolyl 4-hydroxylase subunit beta | P4HB | CLCRP1, DSI, ERBA2L, GIT, P4Hbeta, PDI, PDIA1, PHDB, PO4DB, PO4HB, PROHB | -8.5kb | Sense | Chr. - 81843158 - 81860668 on Build GRCh38 |
Downstream | Aly/REF export factor | ALYREF | ALY, ALY/REF, BEF, REF, THOC4 | 18.2kb | Sense | Chr. - 81887835 - 81891622 on Build GRCh38 |