Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TTTY14
- Entrez Gene ID
-
83869
- Gene Name
- testis-specific transcript, Y-linked 14 (non-protein coding)
- Gene Aliases
-
CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14
- Chromosome Location
-
Chr.Y: 18872501 - 19077547 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125736.1 |
|
3 - 4 |
258 |
130 |
GenBank mRNA |
HY121109.1 |
- |
3 - 4 |
251 |
130 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
HY130111.1 |
- |
3 - 4 |
251 |
|
Chr. Y - 18872737 - 19075996 [-] on Build GRCh38 |
130 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
heat shock transcription factor, Y-linked 2 |
HSFY2 |
HSF2L, HSFY |
-103.3kb
|
Sense |
Chr. - 18731440 - 18790733 on Build GRCh38 |
Downstream |
BCL6 corepressor pseudogene 1 |
BCORP1 |
BCORL2 |
542.9kb
|
Sense |
Chr. - 19455431 - 19503153 on Build GRCh38 |
View Full Product Details