Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINC01612
Entrez Gene ID
101928223
Gene Name
long intergenic non-protein coding RNA 1612
Gene Aliases
-
Chromosome Location
Chr.4: 170226591 - 170283723 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_125889.1 2 - 3 280 102
GenBank mRNA CB163075.1 - 2 - 3 272 102


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CK823081.1 - 2 - 3 261 Chr. 4 - 170226610 - 170283079 [+] on Build GRCh38 102
GenBankmRNA BF593281.1 - 1 - 2 Chr. 4 - 170273919 - 170283034 [-] on Build GRCh38 102
GenBankmRNA BF029192.1 - 2 - 3 259 Chr. 4 - 170226609 - 170283039 [+] on Build GRCh38 102
GenBankmRNA AW271746.1 - 1 - 2 Chr. 4 - 170226713 - 170283033 [-] on Build GRCh38 102
GenBankmRNA BQ101048.1 - 2 - 3 261 Chr. 4 - 170226610 - 170283061 [+] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream aminoadipate aminotransferase AADAT KAT2, KATII, KYAT2 -184.9kb Anti-sense Chr. - 170060222 - 170091699 on Build GRCh38
Downstream heat shock protein 90 alpha family class A member 6, pseudogene HSP90AA6P HSP90Af 321.0kb Anti-sense Chr. - 170604715 - 170605458 on Build GRCh38

View Full Product Details