Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPIN3
Entrez Gene ID
64900
Gene Name
lipin 3
Gene Aliases
LIPN3L, SMP2
Chromosome Location
Chr.20: 41340779 - 41360585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_126051.1 17 - 18 2273 71
XM_011528996.1 XP_011527298.1 17 - 18 2521 71
XM_011528997.2 XP_011527299.1 18 - 19 2727 71
XM_011528998.2 XP_011527300.1 17 - 18 2544 71
XM_011528999.2 XP_011527301.1 18 - 19 2674 71
XM_011529000.1 XP_011527302.1 17 - 18 2518 71
XM_011529002.2 XP_011527304.1 16 - 17 2323 71
XM_011529004.1 XP_011527306.1 18 - 19 2412 71


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX108700.1 - 3 - 4 243 Chr. 20 - 41357111 - 41358763 [+] on Build GRCh38 71
GenBankmRNA AI208765.1 - 3 - 4 Chr. 20 - 41357366 - 41358829 [-] on Build GRCh38 71


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream zinc fingers and homeoboxes 3 ZHX3 TIX1 -40.3kb Anti-sense Chr. - 41178449 - 41317606 on Build GRCh38
Downstream elastin microfibril interfacer 3 EMILIN3 C20orf130, EMILIN5, dJ620E11.4 1.6kb Anti-sense Chr. - 41359966 - 41366858 on Build GRCh38

View Full Product Details