Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARHGAP31-AS1
- Entrez Gene ID
-
100874246
- Gene Name
- ARHGAP31 antisense RNA 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 119314293 - 119322760 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046748.1 |
|
1 - 2 |
148 |
111 |
GenBank mRNA |
DA269443.1 |
- |
1 - 2 |
148 |
111 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
Rho GTPase activating protein 31 |
ARHGAP31 |
AOS1, CDGAP |
|
Anti-sense |
Chr. - 119294289 - 119419476 on Build GRCh38 |
Downstream |
transmembrane protein 39A |
TMEM39A |
|
106.2kb
|
Sense |
Chr. - 119428960 - 119463682 on Build GRCh38 |
View Full Product Details