Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C9orf92
- Entrez Gene ID
-
100129385
- Gene Name
- chromosome 9 open reading frame 92
- Gene Aliases
-
Em:AL513424.1
- Chromosome Location
-
Chr.9: 16203935 - 16276313 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073471.1 |
|
1 - 2 |
86 |
67 |
GenBank mRNA |
AW779702.1 |
- |
3 - 4 |
|
67 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW295211.1 |
- |
3 - 4 |
|
|
Chr. 9 - 16203935 - 16276308 [+] on Build GRCh38 |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
coiled-coil domain containing 171 |
CCDC171 |
C9orf93, bA536D16.1, bA778P13.1 |
-243.9kb
|
Anti-sense |
Chr. - 15552874 - 16062431 on Build GRCh38 |
Downstream |
basonuclin 2 |
BNC2 |
BSN2 |
133.2kb
|
Sense |
Chr. - 16409503 - 16870838 on Build GRCh38 |
View Full Product Details