Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC441242
- Entrez Gene ID
-
441242
- Gene Name
- uncharacterized LOC441242
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 65647010 - 65770810 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103527.2 |
|
3 - 4 |
268 |
74 |
GenBank mRNA |
CN483231.1 |
- |
3 - 4 |
259 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
integrator complex subunit 4-like protein 2 |
LOC101929322 |
|
-159.3kb
|
Sense |
Chr. - 65492233 - 65555065 on Build GRCh38 |
Downstream |
chaperonin containing TCP1 subunit 6 pseudogene 1 |
CCT6P1 |
CCT6-5P, CCT6AP1 |
24.2kb
|
Anti-sense |
Chr. - 65751105 - 65763675 on Build GRCh38 |
View Full Product Details