Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ADORA2A
- Entrez Gene ID
-
135
- Gene Name
- adenosine A2a receptor
- Gene Aliases
-
A2aR, ADORA2, RDC8
- Chromosome Location
-
Chr.22: 24423597 - 24442360 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103544.1 |
|
1 - 2 |
167 |
78 |
GenBank mRNA |
AK301420.1 |
- |
1 - 2 |
102 |
78 |
DA983859.1 |
- |
1 - 2 |
102 |
78 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
HY088145.1 |
- |
1 - 2 |
131 |
|
Chr. 22 - 24427598 - 24440884 [+] on Build GRCh38 |
78 |
GenBankmRNA |
BM917693.1 |
- |
|
87 |
|
Chr. - - [-] on Build |
78 |
GenBankmRNA |
HY091025.1 |
- |
1 - 2 |
131 |
|
Chr. 22 - 24427598 - 24440958 [+] on Build GRCh38 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SPECC1L-ADORA2A readthrough (NMD candidate) |
SPECC1L-ADORA2A |
|
|
Sense |
Chr. - 24270817 - 24442360 on Build GRCh38 |
Downstream |
beta-ureidopropionase 1 |
UPB1 |
BUP1 |
52.1kb
|
Sense |
Chr. - 24495060 - 24528681 on Build GRCh38 |
View Full Product Details