The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_103764.1 | 1 - 2 | 179 | 92 | |
GenBank mRNA | AK092451.1 | - | 1 - 2 | 179 | 92 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | enoyl-CoA delta isomerase 2 | ECI2 | ACBD2, DRS-1, DRS1, HCA88, PECI, dJ1013A10.3 | -475.0 | Anti-sense | Chr. - 4115693 - 4135597 on Build GRCh38 |
Downstream | uncharacterized LOC102724096 | LOC102724096 | 48.9kb | Anti-sense | Chr. - 4186320 - 4190029 on Build GRCh38 |