Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GABRB3
- Entrez Gene ID
-
2562
- Gene Name
- gamma-aminobutyric acid type A receptor beta3 subunit
- Gene Aliases
-
ECA5
- Chromosome Location
-
Chr.15: 26543546 - 26773788 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103801.1 |
|
3 - 4 |
520 |
101 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI832119.1 |
- |
3 - 4 |
524 |
|
Chr. 15 - 26624435 - 26629178 [-] on Build GRCh38 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 929 |
LINC00929 |
|
-491.2kb
|
Anti-sense |
Chr. - 26115813 - 26133037 on Build GRCh38 |
Downstream |
gamma-aminobutyric acid type A receptor alpha5 subunit |
GABRA5 |
|
241.7kb
|
Anti-sense |
Chr. - 26866719 - 26949210 on Build GRCh38 |
View Full Product Details