Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC00676
- Entrez Gene ID
-
101409253
- Gene Name
- long intergenic non-protein coding RNA 676
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 109728274 - 109730034 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103846.1 |
|
1 - 2 |
276 |
81 |
GenBank mRNA |
AW816149.1 |
- |
1 - 2 |
289 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 399 |
LINC00399 |
TCONS_00021596 |
-326.6kb
|
Sense |
Chr. - 109400696 - 109401641 on Build GRCh38 |
Downstream |
insulin receptor substrate 2 |
IRS2 |
IRS-2 |
24.6kb
|
Anti-sense |
Chr. - 109753837 - 109786567 on Build GRCh38 |
View Full Product Details