Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC101060553
- Entrez Gene ID
-
101060553
- Gene Name
- uncharacterized LOC101060553
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 111148197 - 111153577 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120398.1 |
|
1 - 1 |
1518 |
144 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA123006.1 |
- |
1 |
232 |
|
Chr. 13 - 111151722 - 111152290 [-] on Build GRCh38 |
144 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ARHGEF7 antisense RNA 1 |
ARHGEF7-AS1 |
|
-1.5kb
|
Sense |
Chr. - 111144305 - 111148565 on Build GRCh38 |
Downstream |
testis expressed 29 |
TEX29 |
C13orf16, bA474D23.1 |
162.6kb
|
Anti-sense |
Chr. - 111305838 - 111344248 on Build GRCh38 |
View Full Product Details