Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLF4
Entrez Gene ID
9314
Gene Name
Kruppel like factor 4
Gene Aliases
EZF, GKLF
Chromosome Location
Chr.9: 107484852 - 107489720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314052.1 NP_001300981.1 1 - 2 553 93
NM_004235.5 NP_004226.3 1 - 2 553 93
GenBank mRNA AF022184.1 - 1 - 2 404 93
AF105036.1 - 1 - 2 307 93
AK313489.1 - 1 - 2 142 93
BC029923.1 - 1 - 2 118 93
HF546201.1 - 1 - 2 234 93
HF546202.1 - 1 - 2 234 93
HF546213.1 - 1 - 2 234 93
U70663.1 - 1 - 2 481 93

View Full Product Details