The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006172.3 | NP_006163.1 | 3 - 3 | 796 | 91 |
GenBank mRNA | AA779538.1 | - | 1 - 1 | 91 | |
BC005893.1 | - | 3 - 3 | 796 | 91 |