The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000181.3 | NP_000172.2 | 12 - 12 | 2135 | 130 |
NM_001284290.1 | NP_001271219.1 | 10 - 10 | 1697 | 130 | |
NM_001293104.1 | NP_001280033.1 | 11 - 11 | 1950 | 130 | |
NM_001293105.1 | NP_001280034.1 | 10 - 10 | 1807 | 130 | |
NR_120531.1 | 11 - 11 | 2050 | 130 | ||
XM_005250297.3 | XP_005250354.1 | 11 - 11 | 1977 | 130 | |
XM_011516113.1 | XP_011514415.1 | 11 - 11 | 1879 | 130 | |
XM_011516114.1 | XP_011514416.1 | 11 - 11 | 1807 | 130 | |
XM_017012091.1 | XP_016867580.1 | 10 - 10 | 1726 | 130 | |
XM_017012092.1 | XP_016867581.1 | 10 - 10 | 1713 | 130 | |
XM_017012093.1 | XP_016867582.1 | 10 - 10 | 1654 | 130 | |
GenBank mRNA | AK096764.1 | - | 4 - 4 | 2393 | 130 |
AK223406.1 | - | 12 - 12 | 2034 | 130 | |
AK303819.1 | - | 10 - 10 | 1594 | 130 | |
BC014142.2 | - | 12 - 12 | 2005 | 130 | |
BM719712.1 | - | 3 - 3 | 388 | 130 | |
CD014092.1 | - | 11 - 11 | 1832 | 130 | |
CD014093.1 | - | 10 - 10 | 1690 | 130 | |
CD014094.1 | - | 11 - 11 | 1932 | 130 | |
M15182.1 | - | 12 - 12 | 2030 | 130 |