Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
STRC | NM_153700.2 | NP_714544.1 | ||
XM_011521278.2 | XP_011519580.1 | |||
XM_017021954.1 | XP_016877443.1 | |||
XM_017021955.1 | XP_016877444.1 | |||
XM_017021956.1 | XP_016877445.1 | |||
XM_017021957.1 | XP_016877446.1 | |||
XM_017021958.1 | XP_016877447.1 | |||
XM_017021959.1 | XP_016877448.1 | |||
XM_017021962.1 | XP_016877451.1 | |||
AK090757.1 | ||||
AK124582.1 | ||||
AK126721.1 | ||||
BC129931.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832988 | Chr15:43512443 - 43684664 on Build GRCh38 | Loss | STRC MAP1A CKMT1B PPIP5K1 CATSPER2 |
dgv733e212 | Chr15:43596009 - 43703698 on Build GRCh38 | Gain | STRC CKMT1B CATSPER2 STRCP1 CKMT1A |
nsv437799 | Chr15:43491802 - 43697320 on Build GRCh38 | Loss | STRC TP53BP1 MAP1A CKMT1B PPIP5K1 CATSPER2 CKMT1A |
nsv9250 | Chr15:43600811 - 43604249 on Build GRCh38 | Loss | STRC |
nsv433293 | Chr15:43603445 - 43647444 on Build GRCh38 | Loss | STRC CATSPER2 |
dgv2584n100 | Chr15:43600874 - 43703678 on Build GRCh38 | Gain+Loss | STRC CATSPER2 STRCP1 CKMT1A |
nsv1514 | Chr15:43567062 - 43704382 on Build GRCh38 | Deletion | STRC CKMT1B PPIP5K1 CATSPER2 STRCP1 CKMT1A |
nsv1070270 | Chr15:43577801 - 43651202 on Build GRCh38 | Deletion | STRC CKMT1B PPIP5K1 CATSPER2 |
nsv569243 | Chr15:43599314 - 43646090 on Build GRCh38 | Gain | STRC CKMT1B CATSPER2 |
esv3892682 | Chr15:43596778 - 43647444 on Build GRCh38 | Loss | STRC CKMT1B CATSPER2 |
nsv509569 | Chr15:43552272 - 43621659 on Build GRCh38 | Insertion | STRC CKMT1B PPIP5K1 |
esv33424 | Chr15:43596757 - 43703025 on Build GRCh38 | Loss | STRC CKMT1B CATSPER2 STRCP1 CKMT1A |
nsv569247 | Chr15:43601620 - 43647444 on Build GRCh38 | Loss | STRC CATSPER2 |
nsv1160289 | Chr15:43553305 - 43684220 on Build GRCh38 | Deletion | STRC CKMT1B PPIP5K1 CATSPER2 |
nsv428304 | Chr15:43530960 - 43716382 on Build GRCh38 | Loss | STRC MAP1A CKMT1B PPIP5K1 CATSPER2 STRCP1 CKMT1A |
esv2749626 | Chr15:43596837 - 43696748 on Build GRCh38 | Deletion | STRC CKMT1B CATSPER2 CKMT1A |
esv3569487 | Chr15:43559201 - 43684983 on Build GRCh38 | Gain | STRC CKMT1B PPIP5K1 CATSPER2 |
esv3569520 | Chr15:43596064 - 43762536 on Build GRCh38 | Gain | STRC CKMT1B CATSPER2 STRCP1 PDIA3 CKMT1A CATSPER2P1 |
dgv732e212 | Chr15:43576373 - 43703698 on Build GRCh38 | Loss | STRC CKMT1B PPIP5K1 CATSPER2 STRCP1 CKMT1A |
esv24113 | Chr15:43559946 - 43696443 on Build GRCh38 | Gain+Loss | STRC CKMT1B PPIP5K1 CATSPER2 CKMT1A |
dgv45e203 | Chr15:43542120 - 43732292 on Build GRCh38 | Gain+Loss | STRC CKMT1B PPIP5K1 CATSPER2 STRCP1 CKMT1A |
esv2421427 | Chr15:43592920 - 43696355 on Build GRCh38 | Deletion | STRC CKMT1B CATSPER2 CKMT1A |
dgv2583n100 | Chr15:43553212 - 43698816 on Build GRCh38 | Loss | STRC CKMT1B PPIP5K1 CATSPER2 CKMT1A |
nsv1039949 | Chr15:43539681 - 43700448 on Build GRCh38 | Gain | STRC CKMT1B PPIP5K1 CATSPER2 STRCP1 CKMT1A |
nsv974568 | Chr15:43559147 - 43618896 on Build GRCh38 | Duplication | STRC CKMT1B PPIP5K1 |
esv3636291 | Chr15:43598985 - 43609178 on Build GRCh38 | Loss | STRC CKMT1B |
nsv524910 | Chr15:43544280 - 43647444 on Build GRCh38 | Loss | STRC CKMT1B PPIP5K1 CATSPER2 |