Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
YWHAQ | NM_006826.3 | NP_006817.1 | ||
AF070556.1 | AAC28640.1 | |||
AK297624.1 | ||||
AK302208.1 | ||||
AK309462.1 | ||||
AK316158.1 | ||||
BC050601.1 | AAH50601.1 | |||
BC056867.1 | AAH56867.1 | |||
BC093019.1 | AAH93019.1 | |||
BT020014.1 | AAV38817.1 | |||
X56468.1 | CAA39840.1 | |||
X57347.1 | CAA40622.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv580915 | Chr2:9168019 - 9704049 on Build GRCh38 | Gain | IAH1 ADAM17 YWHAQ ASAP2 LOC105373418 LOC101926982 CPSF3 ITGB1BP1 |