The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173699.3 | NP_775970.1 | 2 - 3 | 1249 | 105 |
GenBank mRNA | AK057361.1 | - | 2 - 3 | 1194 | 105 |
BC029525.1 | - | 2 - 3 | 1249 | 105 |