The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152739.3 | NP_689952.1 | 1 - 2 | 654 | 145 |
GenBank mRNA | BC006537.2 | - | 1 - 2 | 634 | 145 |
BC010023.2 | - | 1 - 2 | 634 | 145 | |
BT006990.1 | - | 1 - 2 | 581 | 145 | |
U82759.1 | - | 1 - 2 | 609 | 145 |