Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2D6
Entrez Gene ID
1565
Gene Name
cytochrome P450 family 2 subfamily D member 6
Gene Aliases
CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1
Chromosome Location
Chr.22: 42125531 - 42130881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000106.5 NP_000097.3 3 - 4 594 156
XM_011529966.2 XP_011528268.1 3 - 4 588 156
XM_011529968.2 XP_011528270.1 3 - 4 588 156
XM_011529972.2 XP_011528274.1 3 - 4 588 156
GenBank mRNA BC075023.2 - 3 - 4 534 156
BC075024.2 - 3 - 4 534 156
M20403.1 - 3 - 4 504 156
X08006.1 - 3 - 4 504 156

View Full Product Details