Product Details

Assay Reference Genome
Location

Chr.7:81734722 on build GRCh38
Cytoband
7q21.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
3082
Gene Symbol
HGF
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
Location
Chr.7:81699006-81770438 on build GRCh38
Assay Gene Location
Within Intron 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
HGF NM_000601.5 NP_000592.3
NM_001010932.2 NP_001010932.1
XM_006715956.2 XP_006716019.1
XM_011516115.2 XP_011514417.1
AK291674.1
AK313694.1
BC130284.1
BC130286.1
M29145.1 AAA52650.1
M55379.1
M60718.1 AAA52648.1
M73239.1 AAA64239.1
M73240.1 AAA64297.1
X16323.1 CAA34387.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1019753 Chr7:81692852 - 82052256 on Build GRCh38 Gain CACNA2D1 HGF LOC101927356

View Full Product Details