Product Details

Assay Reference Genome
Location

Chr.7:81703989 on build GRCh38
Cytoband
7q21.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
3082
Gene Symbol
HGF
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
Location
Chr.7:81699006-81770438 on build GRCh38
Assay Gene Location
Within Intron 19
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
HGF NM_000601.5 NP_000592.3
NM_001010932.2 NP_001010932.1
XM_006715956.2 XP_006716019.1
XM_011516115.2 XP_011514417.1
AK291674.1
AK313694.1
BC130284.1
BC130286.1
M29145.1 AAA52650.1
M55379.1
M60718.1 AAA52648.1
M73239.1 AAA64239.1
M73240.1 AAA64297.1
X16323.1 CAA34387.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3576398 Chr7:81685626 - 81732874 on Build GRCh38 Gain HGF LOC100128317
nsv1019753 Chr7:81692852 - 82052256 on Build GRCh38 Gain HGF LOC101927356 CACNA2D1

View Full Product Details