Product Details

Assay Reference Genome
Location

Chr.7:96162814 on build GRCh38
Cytoband
7q21.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
10165
Gene Symbol
SLC25A13
Gene Name
solute carrier family 25 member 13
Gene Aliases
ARALAR2, CITRIN, CTLN2
Location
Chr.7:96120220-96322147 on build GRCh38
Assay Gene Location
Within Intron 16
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SLC25A13 NM_001160210.1 NP_001153682.1
NM_014251.2 NP_055066.1
NR_027662.1
XM_006715831.3 XP_006715894.1
XM_011515727.2 XP_011514029.1
XM_017011663.1 XP_016867152.1
XM_017011664.1 XP_016867153.1
XM_017011665.1 XP_016867154.1
AF118838.1 AAD38501.1
AJ496569.1 CAD43091.1
AK000766.1
AK025227.1
AK025779.1
AK222864.1 BAD96584.1
AK294629.1
BC006566.2 AAH06566.1
Y17571.1 CAB62206.1

View Full Product Details