Product Details

Assay Reference Genome
Location

Chr.7:30964863 on build GRCh38
Cytoband
7p14.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2692
Gene Symbol
GHRHR
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
Location
Chr.7:30964021-30979531 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GHRHR NM_000823.3 NP_000814.2
AY557192.1 AAS59864.1
L01406.1 AAA35890.1
L09237.1 AAA58619.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv6316n100 Chr7:30609596 - 31145482 on Build GRCh38 Gain FAM188B INMT ADCYAP1R1 CRHR2 GHRHR AQP1 GARS INMT-FAM188B
nsv527038 Chr7:30617851 - 31122331 on Build GRCh38 Gain FAM188B INMT ADCYAP1R1 CRHR2 GHRHR AQP1 GARS INMT-FAM188B
esv2761316 Chr7:30615466 - 31127062 on Build GRCh38 Gain FAM188B INMT ADCYAP1R1 CRHR2 GHRHR AQP1 GARS INMT-FAM188B
nsv436557 Chr7:30288062 - 32730902 on Build GRCh38 Deletion DPY19L1P1 INMT PPP1R17 GHRHR GARS INMT-FAM188B DKFZP586I1420 NOD1 LSM5 GGCT MIR550A1 PDE1C ADCYAP1R1 CRHR2 ZNRF2P1 LOC401320 LOC100130673 NEUROD6 CCDC129 FAM188B MIR550B1 AQP1 LINC01176 ZNRF2 AVL9 LOC101928268

View Full Product Details