Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CLN8 | NM_018941.3 | NP_061764.2 | ||
XM_005266021.3 | XP_005266078.1 | |||
XM_005266022.1 | XP_005266079.1 | |||
XM_005266023.1 | XP_005266080.1 | |||
XM_011534745.1 | XP_011533047.1 | |||
XM_011534746.2 | XP_011533048.1 | |||
XM_011534747.1 | 3 | 1553 | XP_011533049.1 | |
AF123757.1 | AAF13115.1 | |||
AF123758.1 | AAF13116.1 | |||
AF123759.1 | AAF13117.1 | |||
AF123760.1 | AAF13118.1 | |||
AF123761.1 | AAF13119.1 | |||
AK312486.1 | ||||
BC007725.2 | AAH07725.1 | |||
BT007049.1 | AAP35698.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv465274 | Chr8:1736898 - 1780126 on Build GRCh38 | Loss | LOC101927752 CLN8 |
nsv1024325 | Chr8:651043 - 3203837 on Build GRCh38 | Gain | LOC105377781 CSMD1 KBTBD11-OT1 LOC286083 LOC100131395 DLGAP2 LOC105377777 LOC101928058 LOC101927752 LOC401442 MYOM2 MIR3674 CLN8 LOC101927815 ERICH1-AS1 KBTBD11 ARHGEF10 LOC105377774 MIR596 LOC105377776 ERICH1 LOC105377792 MIR7160 DLGAP2-AS1 |
esv3891267 | Chr8:1755403 - 2343278 on Build GRCh38 | Gain | LOC105377781 KBTBD11 KBTBD11-OT1 LOC100131395 ARHGEF10 MIR596 LOC101928058 LOC101927752 MYOM2 MIR3674 MIR7160 CLN8 |
esv3615841 | Chr8:1705527 - 1822229 on Build GRCh38 | Loss | DLGAP2 MIR596 LOC101927752 MIR3674 CLN8 |
dgv11822n54 | Chr8:1748920 - 1829405 on Build GRCh38 | Loss | ARHGEF10 MIR596 LOC101927752 MIR3674 CLN8 |
nsv609527 | Chr8:1733567 - 1805331 on Build GRCh38 | Loss | LOC101927752 MIR3674 CLN8 |
esv3576504 | Chr8:951237 - 1906751 on Build GRCh38 | Gain | ERICH1-AS1 LOC286083 ARHGEF10 DLGAP2 MIR596 LOC105377776 LOC101927752 MIR3674 DLGAP2-AS1 CLN8 |
nsv609531 | Chr8:1768870 - 2267169 on Build GRCh38 | Gain | LOC105377781 KBTBD11 KBTBD11-OT1 LOC100131395 ARHGEF10 MIR596 LOC101928058 MYOM2 MIR3674 MIR7160 CLN8 |
nsv509244 | Chr8:1756513 - 1850704 on Build GRCh38 | Insertion | ARHGEF10 MIR596 LOC101927752 MIR3674 CLN8 |
nsv1025591 | Chr8:1755786 - 2335806 on Build GRCh38 | Gain | LOC105377781 KBTBD11 KBTBD11-OT1 LOC100131395 ARHGEF10 MIR596 LOC101928058 LOC101927752 MYOM2 MIR3674 MIR7160 CLN8 |
esv3615846 | Chr8:1735209 - 1816717 on Build GRCh38 | Loss | LOC101927752 MIR3674 CLN8 |