Product Details

Assay Reference Genome
Location

Chr.10:93769768 on build GRCh38
Cytoband
10q23.33
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
9211
Gene Symbol
LGI1
Gene Name
leucine rich glioma inactivated 1
Gene Aliases
ADLTE, ADPAEF, ADPEAF, EPITEMPIN, EPT, ETL1, IB1099
Location
Chr.10:93757770-93798174 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LGI1 NM_001308275.1 NP_001295204.1
NM_001308276.1 NP_001295205.1
NM_005097.3 NP_005088.1
NR_131777.1
XM_017016911.1 XP_016872400.1
XM_017016912.1 XP_016872401.1
AF055636.1 AAC99316.1
AF473548.1 AAM22074.1
AK289706.1
AK303956.1
AK309416.1
AY358885.1 AAQ89244.1
BC022500.1 AAH22500.1
KU178468.1
KU178469.1
KU178470.1
KU178471.1

View Full Product Details