Product Details

Assay Reference Genome
Location

Chr.10:6024036 on build GRCh38
Cytoband
10p15.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
3559
Gene Symbol
IL2RA
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
Location
Chr.10:6010694-6062370 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
IL2RA NM_000417.2 NP_000408.1
NM_001308242.1 NP_001295171.1
NM_001308243.1 NP_001295172.1
AF008556.1 AAB63458.1
AK223313.1 BAD97033.1
AK313847.1
CR996090.1
K03122.1 AAB59535.1
M14098.1 AAA59142.1
X01057.1 CAA25525.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv525927 Chr10:6017935 - 6036116 on Build GRCh38 Loss IL2RA
dgv1e49 Chr10:5684858 - 6183224 on Build GRCh38 Loss MIR3155A PFKFB3 RBM17 FAM208B ANKRD16 GDI2 FBXO18 IL15RA MIR3155B IL2RA

View Full Product Details