Product Details

Assay Reference Genome
Location

Chr.11:113392511 on build GRCh38
Cytoband
11q23.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
255239
Gene Symbol
ANKK1
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
PKK2
Location
Chr.11:113386014-113400418 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ANKK1 NM_178510.1 NP_848605.1
XM_011542736.2 XP_011541038.1
XM_011542737.2 XP_011541039.1
XM_011542738.2 XP_011541040.1
XM_017017475.1 XP_016872964.1
AF487542.1 AAQ09005.1
AJ541797.2 CAD62569.2

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv483080 Chr11:113216072 - 113400474 on Build GRCh38 Loss NCAM1-AS1 NCAM1 TTC12 ANKK1
nsv1040152 Chr11:113387343 - 113415194 on Build GRCh38 Loss DRD2 ANKK1
nsv556444 Chr11:113318201 - 113512672 on Build GRCh38 Gain TTC12 DRD2 MIR4301 ANKK1

View Full Product Details