Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
RNASEH2B | NM_001142279.2 | NP_001135751.1 | ||
NM_024570.3 | NP_078846.2 | |||
XM_006719867.3 | XP_006719930.1 | |||
XM_011535229.1 | XP_011533531.1 | |||
XM_011535230.2 | XP_011533532.1 | |||
XM_011535231.2 | XP_011533533.1 | |||
XM_011535233.2 | XP_011533535.1 | |||
XM_017020747.1 | XP_016876236.1 | |||
AK021774.1 | BAB13892.1 | |||
AK223340.1 | BAD97060.1 | |||
AY764036.1 | AAX13343.1 | |||
BC001397.2 | AAH01397.1 | |||
BC005088.1 | ||||
BC007332.2 | AAH07332.1 | |||
BC010174.2 | AAH10174.1 | |||
BC036744.1 | AAH36744.1 | |||
BX382357.2 | ||||
DB221997.1 | ||||
DT216141.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv561618 | Chr13:49907577 - 51521449 on Build GRCh38 | Loss | RNASEH2B-AS1 DLEU7-AS1 MIR16-1 DLEU7 INTS6 KCNRG ST13P4 RNASEH2B DLEU2 GUCY1B2 DLEU1 SPRYD7 FAM124A MIR5693 LINC00371 TRIM13 MIR15A INTS6-AS1 DLEU1-AS1 MIR3613 SERPINE3 |