Product Details

Assay Reference Genome
Location

Chr.15:58429189 on build GRCh38
Cytoband
15q21.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
3990
Gene Symbol
LIPC
Gene Name
lipase C, hepatic type
Gene Aliases
HDLCQ12, HL, HTGL, LIPH
Location
Chr.15:58410754-58568952 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LIPC XM_005254372.1 XP_005254429.1
AK290062.1
AK292631.1
BC146659.1
Entrez Gene ID
101928635
Gene Symbol
LOC101928635
Gene Name
uncharacterized LOC101928635
Gene Aliases
Location
Chr.15:58106609-58431770 on build GRCh38
Assay Gene Location
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LOC101928635

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv569598 Chr15:58260407 - 58560264 on Build GRCh38 Loss LIPC LIPC-AS1 LOC101928668 LOC101928635
nsv1047048 Chr15:58409201 - 58492601 on Build GRCh38 Loss LIPC LIPC-AS1 LOC101928635

View Full Product Details