Product Details

Assay Reference Genome
Location

Chr.17:28403426 on build GRCh38
Cytoband
17q11.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
113235
Gene Symbol
SLC46A1
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
Location
Chr.17:28394642-28406630 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SLC46A1 NM_001242366.2 NP_001229295.1
NM_080669.5 NP_542400.2
XM_005277786.3 XP_005277843.1
XM_017024110.1 XP_016879599.1
AK054669.1 BAB70789.1
AK074161.1 BAB84987.1
AK097194.1 1 1510
AL832613.2 CAD89945.1
BC010691.1 AAH10691.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1062056 Chr17:28376191 - 28452023 on Build GRCh38 Gain SLC46A1 SARM1
nsv1056839 Chr17:28348199 - 28428188 on Build GRCh38 Gain SEBOX MIR4723 TMEM199 SLC46A1 SARM1 POLDIP2 VTN
nsv574653 Chr17:28227749 - 28501479 on Build GRCh38 Gain RPS7P1 MIR4723 KRT18P55 TMEM97 TMEM199 VTN IFT20 PYY2 PPY2P LOC105371712 LOC101926984 SEBOX SLC13A2 LOC105371930 TNFAIP1 SLC46A1 SARM1 POLDIP2

View Full Product Details