Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
EDARADD | NM_080738.3 | NP_542776.1 | ||
NM_145861.2 | NP_665860.2 | |||
AK096339.1 | ||||
AK290862.1 | ||||
AK291930.1 | ||||
AK314634.1 | ||||
AY028913.1 | AAK40287.1 | |||
AY028914.1 | AAK40288.1 | |||
AY071862.1 | AAL60590.1 | |||
BC128082.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005877 | Chr1:235912335 - 237038508 on Build GRCh38 | Gain | MT1HL1 EDARADD LGALS8 ERO1B LGALS8-AS1 HEATR1 MTR ACTN2 NID1 GPR137B |
esv2763410 | Chr1:236334208 - 236735742 on Build GRCh38 | Gain | EDARADD LGALS8 LGALS8-AS1 HEATR1 ACTN2 |
nsv549394 | Chr1:235876577 - 236629706 on Build GRCh38 | Gain | EDARADD LGALS8 ERO1B LGALS8-AS1 HEATR1 LYST NID1 GPR137B |
nsv1012436 | Chr1:235860657 - 236648100 on Build GRCh38 | Loss | EDARADD LGALS8 ERO1B LGALS8-AS1 HEATR1 LYST NID1 GPR137B |