Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
UVSSA | XM_017008493.1 | XP_016863982.1 | ||
XM_017008496.1 | 1 | 594 | XP_016863985.1 | |
XM_017008497.1 | 1 | 593 | XP_016863986.1 | |
XM_017008499.1 | 1 | 98 | XP_016863988.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv593298 | Chr4:1250479 - 1348733 on Build GRCh38 | Loss | MAEA UVSSA CTBP1-AS2 |
nsv822425 | Chr4:546316 - 1420011 on Build GRCh38 | Loss | RNF212 LOC101928548 DGKQ LOC101928521 TMEM175 CTBP1-AS2 LOC107986211 FGFRL1 SLC26A1 CRIPAK ATP5I LOC105374344 PDE6B TMED11P UVSSA LOC100129917 NKX1-1 SPON2 PCGF3 CPLX1 MAEA LOC100130872 MFSD7 CTBP1-AS MYL5 IDUA CTBP1 GAK |
esv3569247 | Chr4:1340902 - 1353008 on Build GRCh38 | Loss | UVSSA |
nsv593278 | Chr4:1165892 - 1421910 on Build GRCh38 | Loss | MAEA LOC100130872 UVSSA LOC101928548 CTBP1-AS NKX1-1 CTBP1 SPON2 CRIPAK CTBP1-AS2 |
nsv593301 | Chr4:1306289 - 1358897 on Build GRCh38 | Loss | MAEA UVSSA |
nsv593302 | Chr4:1344830 - 1420896 on Build GRCh38 | Loss | UVSSA NKX1-1 CRIPAK |
nsv508988 | Chr4:1190955 - 1358870 on Build GRCh38 | Insertion | MAEA LOC100130872 UVSSA LOC101928548 CTBP1-AS CTBP1 SPON2 CTBP1-AS2 |
nsv593281 | Chr4:1209477 - 1358897 on Build GRCh38 | Loss | MAEA UVSSA CTBP1-AS CTBP1 CTBP1-AS2 |