Product Details
- Assay Reference Genome
Location
Chr.5:78016773 on build GRCh38
- Cytoband
- 5q14.1
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
For this assay, SNP(s) [rs73132861] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Target Gene Details
- Entrez Gene ID
- 8546
- Gene Symbol
- AP3B1
- Gene Name
- adaptor related protein complex 3 beta 1 subunit
- Gene Aliases
- ADTB3, ADTB3A, HPS, HPS2, PE
- Location
- Chr.5:78002326-78294755 on build GRCh38
- Assay Gene Location
- Within Intron 26
View Full Product Details