Product Details

Assay Reference Genome
Location

Chr.8:38415169 on build GRCh38
Cytoband
8p11.23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2260
Gene Symbol
FGFR1
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Location
Chr.8:38411138-38468834 on build GRCh38
Assay Gene Location
Within Intron 18
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FGFR1 NM_001174063.1 NP_001167534.1
NM_001174064.1 NP_001167535.1
NM_001174065.1 NP_001167536.1
NM_001174066.1 NP_001167537.1
NM_001174067.1 NP_001167538.1
NM_015850.3 NP_056934.2
NM_023105.2 NP_075593.1
NM_023106.2 NP_075594.1
NM_023110.2 NP_075598.2
XM_006716303.2 XP_006716366.1
XM_006716304.1 XP_006716367.1
XM_006716306.2 XP_006716369.1
XM_006716307.1 XP_006716370.1
XM_006716309.3 XP_006716372.1
XM_006716310.2 XP_006716373.1
XM_006716311.1 XP_006716374.1
XM_006716312.1 XP_006716375.1
XM_006716313.2 XP_006716376.1
XM_006716314.1 XP_006716377.1
XM_011544443.2 XP_011542745.1
XM_011544444.1 XP_011542746.1
XM_011544445.2 XP_011542747.1
XM_011544446.2 XP_011542748.1
XM_011544447.2 XP_011542749.1
XM_011544448.1 XP_011542750.1
XM_011544449.1 XP_011542751.1
XM_011544450.2 XP_011542752.1
XM_011544451.1 XP_011542753.1
XM_017013219.1 XP_016868708.1
XM_017013220.1 XP_016868709.1
XM_017013221.1 XP_016868710.1
XM_017013222.1 XP_016868711.1
XM_017013223.1 XP_016868712.1
XM_017013224.1 XP_016868713.1
XM_017013225.1 XP_016868714.1
XM_017013226.1 XP_016868715.1
XM_017013227.1 XP_016868716.1
XM_017013228.1 XP_016868717.1
XM_017013229.1 XP_016868718.1
XM_017013230.1 XP_016868719.1
AB208919.1 BAD92156.1
AK222718.1 BAD96438.1
AK291754.1
AK292470.1
AK309947.1
BC015035.1 AAH15035.1
BC018128.1 AAH18128.1
BC091494.1 AAH91494.1
BX649167.1
FJ809917.1
KU178066.1
KU178067.1
KU178068.1
KU178069.1
KU178070.1
M34185.1 AAA35836.1
M34186.1 AAA35837.1
M34641.1 AAA35835.1
M37722.1 AAA75007.1
M60485.1 AAA35840.1
M63887.1 AAA35958.1
M63888.1 AAA35959.1
X51803.1 CAA36101.1
X52833.1 CAA37015.1
X57119.1 CAA40401.1
X57120.1 CAA40402.1
X57121.1 CAA40403.1
X57122.1 CAA40404.1
X66945.1 CAA47375.1
Y00665.1 CAA68679.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951128 Chr8:38411783 - 38418282 on Build GRCh38 Deletion FGFR1

View Full Product Details