Product Details

Assay Reference Genome
Location

Chr.11:46647158 on build GRCh38
Cytoband
11p11.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
9776
Gene Symbol
ATG13
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
Location
Chr.11:46617276-46676019 on build GRCh38
Assay Gene Location
Within Intron 10
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ATG13 NM_001142673.2 NP_001136145.1
NM_001205119.1 NP_001192048.1
NM_001205120.1 NP_001192049.1
NM_001205121.1 NP_001192050.1
NM_001205122.1 NP_001192051.1
NM_014741.4 NP_055556.2
XM_005253262.2 XP_005253319.1
XM_005253263.2 XP_005253320.1
XM_005253265.2 XP_005253322.1
XM_005253266.2 XP_005253323.1
XM_005253268.2 XP_005253325.1
XM_006718394.2 XP_006718457.1
XM_006718395.2 XP_006718458.1
XM_006718396.2 XP_006718459.1
XM_011520493.1 XP_011518795.1
XM_011520496.2 XP_011518798.1
XM_011520499.2 XP_011518801.1
XM_011520500.1 XP_011518802.1
XM_011520501.1 XP_011518803.1
XM_011520502.1 XP_011518804.1
XM_011520503.1 XP_011518805.1
XM_017018599.1 XP_016874088.1
XM_017018600.1 XP_016874089.1
XM_017018601.1 XP_016874090.1
XM_017018602.1 XP_016874091.1
XM_017018603.1 XP_016874092.1
XM_017018604.1 XP_016874093.1
XM_017018605.1 XP_016874094.1
XM_017018606.1 XP_016874095.1
XM_017018607.1 XP_016874096.1
XM_017018608.1 XP_016874097.1
XM_017018609.1 XP_016874098.1
XM_017018610.1 XP_016874099.1
XM_017018611.1 XP_016874100.1
XM_017018612.1 XP_016874101.1
XM_017018613.1 XP_016874102.1
XM_017018614.1 XP_016874103.1
XM_017018615.1 XP_016874104.1
XM_017018616.1 XP_016874105.1
XM_017018617.1 XP_016874106.1
XM_017018618.1 XP_016874107.1
XM_017018619.1 XP_016874108.1
XM_017018620.1 XP_016874109.1
XM_017018621.1 XP_016874110.1
XM_017018622.1 XP_016874111.1
AB014552.1
AK023867.1 BAB14707.1
AK223603.1 BAD97323.1
AK289641.1
AK293109.1
AK294110.1
BC001331.1 AAH01331.1
BC002378.2 AAH02378.1
BC006191.2 AAH06191.1
CD675643.1

View Full Product Details