Product Details

Assay Reference Genome
Location

Chr.3:46860527 on build GRCh38
Cytoband
3p21.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
4634
Gene Symbol
MYL3
Gene Name
myosin light chain 3
Gene Aliases
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl
Location
Chr.3:46857867-46863483 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
MYL3 NM_000258.2 NP_000249.1
AF174483.1 AAF91089.1
AK312044.1
BC009790.1 AAH09790.1
BP316922.1
M24122.1 AAA59895.1
X07373.1 CAA30292.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv508915 Chr3:46814349 - 46913473 on Build GRCh38 Insertion PRSS42 PTH1R MYL3
nsv428417 Chr3:46638962 - 47778557 on Build GRCh38 Loss PRSS50 SNORD13P3 SCAP PTPN23 NBEAL2 NRADDP PRSS42 ELP6 CSPG5 MYL3 ALS2CL PRSS45 KIF9 KLHL18 PRSS46 TMIE SMARCC1 CCDC12 PTH1R PRSS43 KIF9-AS1 SETD2
dgv8308n54 Chr3:46811189 - 46894037 on Build GRCh38 Loss PRSS42 PTH1R MYL3 PRSS43

View Full Product Details