Product Details

Assay Reference Genome
Location

Chr.9:27549432 on build GRCh38
Cytoband
9p21.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
203228
Gene Symbol
C9orf72
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
Location
Chr.9:27546546-27573866 on build GRCh38
Assay Gene Location
Within Intron 10
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
C9orf72 NM_001256054.2 NP_001242983.1
NM_018325.4 NP_060795.1
AK001971.1 BAA92009.1
AK291425.1
AL832229.1
AL832467.1
BC068445.1 AAH68445.1
JN681271.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1015259 Chr9:26347305 - 27643043 on Build GRCh38 Gain IFT74-AS1 IFT74 EQTN PLAA IFNK LINC00032 MOB3B C9orf72 CAAP1 TEK LRRC19

View Full Product Details