Product Details

Assay Reference Genome
Location

Chr.12:13555245 on build GRCh38
Cytoband
12p13.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2904
Gene Symbol
GRIN2B
Gene Name
glutamate ionotropic receptor NMDA type subunit 2B
Gene Aliases
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
Location
Chr.12:13537337-13982012 on build GRCh38
Assay Gene Location
Within Exon 16
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GRIN2B XM_005253351.3 4 9899 XP_005253408.1
XM_011520628.2 14 12965 XP_011518930.1
XM_011520629.2 15 12772 XP_011518931.1
XM_017019219.1 14 12520 XP_016874708.1

View Full Product Details